Assay Details
Gene Symbol: nej | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201646.2 | NP_001188575.1 | 8 - 9 | 7327 | 70 | |
NM_001201647.2 | NP_001188576.1 | 8 - 9 | 7279 | 70 | ||
NM_001272458.1 | NP_001259387.1 | 8 - 9 | 7181 | 70 | ||
NM_079903.4 | NP_524642.2 | 8 - 9 | 7356 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
