Assay Details
Gene Symbol: MED20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273294.1 | NP_001260223.1 | 1 | 771 | 77 | |
NM_079924.3 | NP_524663.2 | 1 | 771 | 77 | ||
GenBank mRNA | BT003473.1 | - | 1 | 767 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
