Assay Details
| Gene Symbol: spag | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079925.3 | NP_524664.1 | 4 - 5 | 1311 | 95 | |
| GenBank mRNA | AJ252086.2 | - | 4 - 5 | 1330 | 95 | |
| BT150413.1 | - | 4 - 5 | 1307 | 95 | ||
More Information
Set Membership: |
|

spag
Genomic Map