Assay Details
| Gene Symbol: Ubc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298405.1 | NP_001285334.1 | 2 - 3 | 452 | 70 | |
| NM_079945.4 | NP_524684.2 | 2 - 3 | 452 | 70 | ||
| GenBank mRNA | AY071319.1 | - | 2 - 3 | 450 | 70 | |
More Information
Set Membership: |
|

Ubc7
Genomic Map