Assay Details
Gene Symbol: Ubc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298405.1 | NP_001285334.1 | 3 - 4 | 714 | 70 | |
NM_079945.4 | NP_524684.2 | 3 - 4 | 714 | 70 | ||
GenBank mRNA | AY071319.1 | - | 3 - 4 | 712 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
