Assay Details
| Gene Symbol: PIG-V | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299579.1 | NP_001286508.1 | 2 | 1588 | 81 | |
| NM_001299580.1 | NP_001286509.1 | 1 | 1648 | 81 | ||
| NM_001299581.1 | NP_001286510.1 | 2 | 1577 | 81 | ||
| GenBank mRNA | BT016020.1 | - | 2 | 1582 | 81 | |
More Information
Set Membership: |
|

PIG-V
Genomic Map