Assay Details
Gene Symbol: Mcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079949.3 | NP_524688.1 | 5 - 6 | 5144 | 101 | |
GenBank mRNA | AY058526.1 | - | 5 - 6 | 5110 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
