Assay Details
| Gene Symbol: AstA-R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079961.4 | NP_524700.1 | 5 - 6 | 1179 | 63 | |
| NM_166982.2 | NP_726877.2 | 4 - 5 | 1130 | 63 | ||
| GenBank mRNA | AF163775.1 | - | 3 - 4 | 644 | 63 | |
| AF220216.1 | - | 5 - 6 | 917 | 63 | ||
| BT150204.1 | - | 5 - 6 | 1182 | 63 | ||
More Information
Set Membership: |
|

AstA-R1
Genomic Map