Assay Details
| Gene Symbol: Hmt4-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258524.1 | NP_001245453.1 | 2 - 3 | 803 | 80 | |
| NM_130497.4 | NP_569853.1 | 2 - 3 | 1383 | 80 | ||
More Information
Set Membership: |
|

Hmt4-20
Genomic Map