Assay Details
| Gene Symbol: Jhedup | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137241.4 | NP_611085.2 | 1 - 2 | 186 | 79 | |
| GenBank mRNA | BT133360.1 | - | 1 - 2 | 186 | 79 | |
More Information
Set Membership: |
|

Jhedup
Genomic Map