Assay Details
Gene Symbol: calypso | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137252.3 | NP_611096.1 | 2 - 3 | 548 | 83 | |
NM_176194.2 | NP_788374.1 | 2 - 3 | 518 | 83 | ||
GenBank mRNA | AY047515.1 | - | 2 - 3 | 517 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
