Assay Details
Gene Symbol: Nox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103866.2 | NP_001097336.1 | 3 - 4 | 1283 | 62 | |
NM_001299556.1 | NP_001286485.1 | 3 - 4 | 1387 | 62 | ||
NM_001299557.1 | NP_001286486.1 | 3 - 4 | 3163 | 62 | ||
NM_137272.2 | NP_611116.2 | 3 - 4 | 1490 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
