Assay Details
Gene Symbol: Nox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103866.2 | NP_001097336.1 | 4 - 5 | 1502 | 85 | |
NM_001299556.1 | NP_001286485.1 | 4 - 5 | 1606 | 85 | ||
NM_001299557.1 | NP_001286486.1 | 4 - 5 | 3382 | 85 | ||
NM_137272.2 | NP_611116.2 | 4 - 5 | 1709 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
