Assay Details
| Gene Symbol: NiPp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137333.4 | NP_611177.1 | 2 - 3 | 332 | 77 | |
| GenBank mRNA | AJ427611.1 | - | 2 - 3 | 335 | 77 | |
| AY069781.1 | - | 2 - 3 | 272 | 77 | ||
More Information
Set Membership: |
|

NiPp1
Genomic Map