Assay Details
Gene Symbol: sub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299619.1 | NP_001286548.1 | 2 - 3 | 1841 | 145 | |
NM_137416.3 | NP_611260.1 | 2 - 3 | 1841 | 145 | ||
GenBank mRNA | AY069597.1 | - | 2 - 3 | 1812 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
