Assay Details
| Gene Symbol: Sema5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169959.2 | NP_001163430.1 | 4 - 5 | 736 | 66 | |
| NM_137449.3 | NP_611293.3 | 4 - 5 | 950 | 66 | ||
| GenBank mRNA | BT009966.1 | - | 4 - 5 | 733 | 66 | |
More Information
Set Membership: |
|

Sema5c
Genomic Map