Assay Details
Gene Symbol: Sema5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169959.2 | NP_001163430.1 | 8 - 9 | 2055 | 71 | |
NM_137449.3 | NP_611293.3 | 8 - 9 | 2269 | 71 | ||
GenBank mRNA | BT009966.1 | - | 8 - 9 | 2052 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
