Assay Details
| Gene Symbol: EMRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137450.2 | NP_611294.1 | 1 | 563 | 110 | |
| GenBank mRNA | AY095085.1 | - | 1 | 550 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMRE| Gene Symbol: EMRE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137450.2 | NP_611294.1 | 1 | 563 | 110 | |
| GenBank mRNA | AY095085.1 | - | 1 | 550 | 111 | |
Set Membership: |
|