Assay Details
| Gene Symbol: Topors | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274154.1 | NP_001261083.1 | 2 - 3 | 1054 | 78 | |
| NM_137544.3 | NP_611388.1 | 1 - 2 | 798 | 78 | ||
| GenBank mRNA | AY122186.1 | - | 1 - 2 | 776 | 78 | |
More Information
Set Membership: |
|

Topors
Genomic Map