Assay Details
| Gene Symbol: mip40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137563.4 | NP_611407.1 | 1 | 849 | 111 | |
| GenBank mRNA | AY069362.1 | - | 1 | 829 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mip40| Gene Symbol: mip40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137563.4 | NP_611407.1 | 1 | 849 | 111 | |
| GenBank mRNA | AY069362.1 | - | 1 | 829 | 111 | |
Set Membership: |
|