Assay Details
Gene Symbol: Rgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274158.1 | NP_001261087.1 | 4 - 5 | 963 | 64 | |
NM_137567.3 | NP_611411.2 | 4 - 5 | 963 | 64 | ||
NM_166341.3 | NP_725875.3 | 11 - 12 | 4460 | 64 | ||
GenBank mRNA | BT010242.1 | - | 4 - 5 | 957 | 64 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
