Assay Details
Gene Symbol: Obp56a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137598.3 | NP_611442.1 | 1 - 2 | 105 | 87 | |
GenBank mRNA | AY071723.1 | - | 1 - 2 | 104 | 87 | |
AY113487.1 | - | 1 - 2 | 103 | 87 | ||
KX531909.1 | - | 1 - 2 | 58 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
