Assay Details
| Gene Symbol: Obp57e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137644.1 | NP_611488.1 | 1 - 2 | 56 | 95 | |
| GenBank mRNA | BT100326.2 | - | 1 - 2 | 56 | 95 | |
| KX531744.1 | - | 1 - 2 | 56 | 95 | ||
More Information
Set Membership: |
|

Obp57e
Genomic Map