Assay Details
Gene Symbol: Nnf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137648.4 | NP_611492.1 | 1 - 2 | 166 | 89 | |
GenBank mRNA | AY071387.1 | - | 1 - 2 | 135 | 89 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
