Assay Details
Gene Symbol: Rcd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169746.2 | NP_001163217.1 | 2 - 3 | 1141 | 84 | |
NM_137653.4 | NP_611497.1 | 2 - 3 | 580 | 84 | ||
GenBank mRNA | AY061284.1 | - | 2 - 3 | 547 | 84 | |
BT003220.1 | - | 2 - 3 | 1137 | 84 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
