Assay Details
| Gene Symbol: Rcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169746.2 | NP_001163217.1 | 3 - 4 | 1390 | 72 | |
| NM_137653.4 | NP_611497.1 | 3 - 4 | 829 | 72 | ||
| GenBank mRNA | AY061284.1 | - | 3 - 4 | 796 | 72 | |
| BT003220.1 | - | 3 - 4 | 1386 | 72 | ||
More Information
Set Membership: |
|

Rcd6
Genomic Map