Assay Details
| Gene Symbol: DMAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137659.5 | NP_611503.1 | 2 - 3 | 626 | 61 | |
| GenBank mRNA | AY058649.1 | - | 2 - 3 | 589 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMAP1| Gene Symbol: DMAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137659.5 | NP_611503.1 | 2 - 3 | 626 | 61 | |
| GenBank mRNA | AY058649.1 | - | 2 - 3 | 589 | 61 | |
Set Membership: |
|