Assay Details
| Gene Symbol: Lapsyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137717.4 | NP_611561.1 | 4 - 5 | 832 | 67 | |
| GenBank mRNA | AY051578.1 | - | 4 - 5 | 815 | 67 | |
More Information
Set Membership: |
|

Lapsyn
Genomic Map