Assay Details
| Gene Symbol: Pde9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272566.1 | NP_001259495.1 | 3 - 4 | 3497 | 58 | |
| NM_001272567.2 | NP_001259496.1 | 4 - 5 | 3632 | 58 | ||
| NM_132606.5 | NP_572834.2 | 4 - 5 | 3632 | 58 | ||
| NM_167340.5 | NP_727644.3 | 4 - 5 | 3632 | 58 | ||
More Information
Set Membership: |
|

Pde9
Genomic Map