Assay Details
| Gene Symbol: Fur2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_167506.3 | NP_727963.1 | 4 - 5 | 1388 | 80 | |
| NM_167507.3 | NP_727964.1 | 4 - 5 | 1488 | 80 | ||
| NM_167508.2 | NP_727965.1 | 4 - 5 | 1422 | 80 | ||
More Information
Set Membership: |
|

Fur2
Genomic Map