Assay Details
Gene Symbol: Mgstl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298578.1 | NP_001285507.1 | 1 - 2 | 524 | 81 | |
NM_167724.3 | NP_728382.1 | 1 - 2 | 164 | 81 | ||
GenBank mRNA | AY071752.1 | - | 1 - 2 | 160 | 81 | |
KX531522.1 | - | 1 - 2 | 130 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
