Assay Details
Gene Symbol: tty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272838.1 | NP_001259767.1 | 4 - 5 | 1781 | 65 | |
NM_001272839.2 | NP_001259768.1 | 4 - 5 | 1719 | 65 | ||
NM_001272840.2 | NP_001259769.1 | 4 - 5 | 1779 | 65 | ||
NM_080712.7 | NP_542443.4 | 4 - 5 | 1781 | 65 | ||
NM_167744.4 | NP_728415.4 | 4 - 5 | 2379 | 65 | ||
GenBank mRNA | AF184227.1 | - | 4 - 5 | 1698 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
