Assay Details
Gene Symbol: Cda4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167788.2 | NP_728468.1 | 1 - 2 | 197 | 88 | |
GenBank mRNA | AY113493.1 | - | 1 - 2 | 196 | 88 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
