Assay Details
| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274254.1 | NP_001261183.1 | 4 - 5 | 2305 | 57 | |
| NM_080382.4 | NP_525121.2 | 4 - 5 | 2585 | 57 | ||
| NM_167789.2 | NP_728470.1 | 4 - 5 | 1943 | 57 | ||
| NM_167790.3 | NP_728471.2 | 4 - 5 | 2305 | 57 | ||
| NM_167791.3 | NP_728472.1 | 4 - 5 | 2110 | 57 | ||
| NM_167792.3 | NP_728473.1 | 5 - 6 | 2295 | 57 | ||
| NM_167793.3 | NP_728474.1 | 2 - 3 | 1248 | 57 | ||
| NM_167794.3 | NP_728475.1 | 2 - 3 | 1298 | 57 | ||
| NM_167795.3 | NP_728476.1 | 3 - 4 | 1615 | 57 | ||
| GenBank mRNA | BT004489.1 | - | 4 - 5 | 2560 | 57 | |
| BT011471.1 | - | 3 - 4 | 1590 | 57 | ||
| BT099676.1 | - | 2 - 3 | 1519 | 57 | ||
| BT133220.1 | - | 4 - 5 | 2188 | 57 | ||
More Information
Set Membership: |
|

Pdk1
Genomic Map