Assay Details
Gene Symbol: E(bx) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169833.2 | NP_001163304.1 | 11 - 12 | 8312 | 111 | |
NM_001274258.1 | NP_001261187.1 | 12 - 13 | 7989 | 111 | ||
NM_001274259.1 | NP_001261188.1 | 12 - 13 | 8633 | 111 | ||
NM_001274260.1 | NP_001261189.1 | 11 - 12 | 7942 | 111 | ||
NM_001274261.1 | NP_001261190.1 | 11 - 12 | 7854 | 111 | ||
NM_167817.3 | NP_728505.1 | 12 - 13 | 8297 | 111 | ||
NM_167819.3 | NP_728507.1 | 11 - 12 | 8357 | 111 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
