Assay Details
Gene Symbol: Atac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144394.2 | NP_001137866.1 | 2 - 3 | 578 | 95 | |
NM_001259602.2 | NP_001246531.1 | 3 - 4 | 680 | 95 | ||
NM_001274275.1 | NP_001261204.1 | 2 - 3 | 578 | 95 | ||
NM_167828.4 | NP_728520.2 | 3 - 4 | 731 | 95 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
