Assay Details
Gene Symbol: MED22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130514.5 | NP_569870.1 | 1 - 2 | 196 | 68 | |
GenBank mRNA | AY071113.1 | - | 1 - 2 | 194 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
