Assay Details
| Gene Symbol: MED22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130514.5 | NP_569870.1 | 2 - 3 | 293 | 72 | |
| GenBank mRNA | AY071113.1 | - | 2 - 3 | 291 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED22| Gene Symbol: MED22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130514.5 | NP_569870.1 | 2 - 3 | 293 | 72 | |
| GenBank mRNA | AY071113.1 | - | 2 - 3 | 291 | 72 | |
Set Membership: |
|