Assay Details
| Gene Symbol: Pi4KIIIalpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272262.2 | NP_001259191.1 | 7 - 8 | 4937 | 57 | |
| NM_130658.3 | NP_570014.3 | 6 - 7 | 4877 | 57 | ||
More Information
Set Membership: |
|

Pi4KIIIalpha
Genomic Map