Assay Details
Gene Symbol: ttm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316628.1 | NP_001303557.1 | 3 - 4 | 1117 | 88 | |
NM_130671.3 | NP_570027.1 | 3 - 4 | 1117 | 88 | ||
GenBank mRNA | AY070535.1 | - | 3 - 4 | 1093 | 88 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
