Assay Details
| Gene Symbol: dnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274208.2 | NP_001261137.1 | 1 - 2 | 637 | 86 | |
| NM_137836.3 | NP_611680.2 | 1 - 2 | 637 | 86 | ||
| GenBank mRNA | AY095032.1 | - | 1 - 2 | 625 | 86 | |
More Information
Set Membership: |
|

dnr1
Genomic Map