Assay Details
Gene Symbol: RpS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299802.1 | NP_001286731.1 | 4 - 5 | 402 | 61 | |
NM_137841.4 | NP_611685.1 | 3 - 4 | 221 | 61 | ||
GenBank mRNA | AY070671.1 | - | 3 - 4 | 227 | 61 | |
KX532104.1 | - | 2 - 3 | 190 | 61 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
