Assay Details
Gene Symbol: bonsai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137847.3 | NP_611691.1 | 2 - 3 | 419 | 68 | |
GenBank mRNA | AY060712.1 | - | 2 - 3 | 388 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
