Assay Details
Gene Symbol: stl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103953.3 | NP_001097423.1 | 2 - 3 | 670 | 65 | |
NM_001259540.1 | NP_001246469.1 | 2 - 3 | 1178 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
