Assay Details
| Gene Symbol: EMC8-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137887.4 | NP_611731.1 | 1 - 2 | 294 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMC8-9| Gene Symbol: EMC8-9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137887.4 | NP_611731.1 | 1 - 2 | 294 | 100 | |
Set Membership: |
|