Assay Details
Gene Symbol: RpL37b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137913.3 | NP_611757.1 | 1 | 340 | 56 | |
GenBank mRNA | BT044298.1 | - | 1 | 321 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
