Assay Details
Gene Symbol: wmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103966.2 | NP_001097436.1 | 3 - 4 | 1216 | 78 | |
NM_137960.3 | NP_611804.1 | 2 - 3 | 1161 | 78 | ||
GenBank mRNA | AY061000.1 | - | 2 - 3 | 1127 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
