Assay Details
| Gene Symbol: fzr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138010.4 | NP_611854.1 | 1 | 1313 | 99 | |
| GenBank mRNA | AY051537.1 | - | 1 | 1284 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

fzr2| Gene Symbol: fzr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138010.4 | NP_611854.1 | 1 | 1313 | 99 | |
| GenBank mRNA | AY051537.1 | - | 1 | 1284 | 99 | |
Set Membership: |
|