Assay Details
Gene Symbol: Unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014545.3 | NP_001014545.3 | 24 - 25 | 7446 | 60 | |
NM_001103970.2 | NP_001097440.1 | 25 - 26 | 7518 | 60 | ||
NM_001299874.1 | NP_001286803.1 | 24 - 25 | 7443 | 60 | ||
NM_001299875.1 | NP_001286804.1 | 21 - 22 | 6594 | 60 | ||
NM_001299876.1 | NP_001286805.1 | 21 - 22 | 4609 | 60 | ||
NM_001299877.1 | NP_001286806.1 | 24 - 25 | 6573 | 60 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
