Assay Details
Gene Symbol: CG13579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274240.2 | NP_001261169.1 | 3 - 4 | 908 | 67 | |
NM_138073.4 | NP_611917.2 | 3 - 4 | 908 | 67 |
Gene Symbol: sta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT029094.1 | - | 892 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT029094.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
